To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Melanoma Antigen Family A3
- Clonality:Monoclonal
- Clone:1A9F3
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:MAGEA3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:CT1.3|MAGEA6|HIP8|MAGE3|HYPD
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Caprylic Acid/Ammonium Sulfate Precipitation
- Cat. no.:10089-878
- Supplier No.:60054-1-IG
Spécifications
A propos de cet article
MAGEA3, also named as MAGE3 CT1.3 and MZ2-D, is a member of the MAGE gene family, which comprises 12 known genes, of which 6 are expressed in tumors. The cellular function of this family is unknown. Many publication cite that "MAGE-3 is a tumor-specific antigen widely expressed in solid and hematologic malignancies, but not in normal cells, with the exception of the male germ cells, placenta, and possibly cells of the developing embryo; therefore, MAGEA3 is an excellent candidate tumor antigen." But this antibody can get target band in Fetal Brain and Testis. Novus' antibody can recognize the target band in Fetal Liver. And the MW is about 35-55kd. MAGEA3 has been described as a cytoplasmic protein.
Western Blot: Human Testis Tissue, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200
Type: Primary
Antigen: MAGEA3
Clonality: Monoclonal
Clone: 1A9F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human