To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Neural Precursor Cell Expressed, Developmentally Down-Regulated 8
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NEDD8
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:NEDD 8|NED8|Neural precursor cell expressed developmentally down regulated gene 8|Neddylin|Ubiquitin like protein Nedd 8|Neural precursor cell expressed developmentally down regulated 8|Ubiquitin like protein Nedd8.|Rub1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:16777-1-AP
Spécifications
A propos de cet article
NEDD8 (neural precursor cell-expressed developmentally down-regulated gene 8) is a small ubiquitin-like protein that shares 60% sequence identity and 80% homology with ubiquitin . NEDD8 is conjugated to substrate proteins in a process known as neddylation. NEDD8 forms a thioester bond with APPBP1-Uba3, the E1 enzyme. Activated NEDD8 then transfers to Ubc12, the E2 enzyme . Ultimately, E2 loaded with NEDD8 binds a substrate protein lysine residue and forms an isopeptide bond by E3 ligase .
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200
Type: Primary
Antigen: NEDD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat