To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Bone Morphogenetic Protein Receptor Type 2
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:BMPR2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:type II activin receptor-like kinase|BMPR-II|PPH1|BRK-3|bone morphogenetic protein receptor type II|BMR2|T-ALK|bone morphogenetic protein receptor type-2|BMP type II receptor|BMPR-2|BMPR3|BMP type-2 receptor|type II receptor for bone morphogenetic protein-4
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Supplier No.:14376-1-AP
Spécifications
A propos de cet article
BMPs (bone morphogenetic protein) are involved in endochondral bone formation and embryogenesis. BMPR2 encodes a member of the (BMP) receptor family of transmembrane serine/threonine kinases. It is a widely expressed receptor, with high mRNA expression during development in many tissues. Dimer of BMPR2 receptors (70-80 kD) forms a complex with a dimer of type I BMPR1(50-55 kD) in signal transduction. This antibody recognizes endogenous levels of total BMPR2 protein, including 115 kD precursor, 75 kD mature form and 150 kD dimer.
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IP: Mouse Brain Tissue, 1:200-1:2000
Type: Primary
Antigen: BMPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat