To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Angiotensinogen
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:AGT
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:Member 8|Clade A|Serpin Peptidase Inhibitor|SERPINA8
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10081-910
- Supplier No.:11992-1-AP
Spécifications
A propos de cet article
Angiotensinogen is a precursor of angiotensin II (Ang II), is expressed and synthesized largely in the liver and is cleaved by the enzyme renin in response to lowered blood pressure. It has a key role in mediating vascular constriction and regulating salt and fluid homeostasis. The resulting product, angiotensin I, is then cleaved by angiotensin converting enzyme (ACE) to generate the physiologically active enzyme angiotensin II. Mutations in this gene are associated with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. Defects in this gene have also been associated with non-familial structural atrial fibrillation, and inflammatory bowel disease.
Western Blot: HepG2 Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200; FC: HeLa Cells, N/A
Type: Primary
Antigen: AGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse