To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Tropomyosin 3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TPM3
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:TM30nm|OK/SW-cl.5|HEL-S-82p|TM-5|TM3|HEL-189|TPMsk3|TM30|CFTD|hscp30|CAPM1|NEM1|TRK|TM5
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10096-226
- Supplier No.:10737-1-AP
Spécifications
A propos de cet article
Tropomyosins are ubiquitous proteins of 35 to 45 kD associated with the actin filaments of myofibrils and stress fibers.
Western Blot: Mouse Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200; IP: HeLa Cells, 1:200-1:1000
Type: Primary
Antigen: TPM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat