Acid Digestion Vessel, General Purpose, Parr Instrument Company
Fournisseur: Parr Instrument
Certificats
A propos de cet article
This is a 45 mL modification of the 23 mL, model 4749 PTFE lined vessel for users who want additional capacity in this popular model
It is made with the same closure as the 23 mL vessel, using a similar thick-walled PTFE cup, 33/4" deep inside, with a broad flanged seal. Safety blow out protection is provided with a rupture disc that will release pressure through an opening in the cover. It has a maximum temperature and pressure rating of 250 °C and vessel 1800 psig
Ordering information: The spanner and holding fixture accessories are required for cap tightening, but are not included with bomb. These accessories must be ordered separately.
Customers Who Bought This Also Bought