Axygen® Sequencing Septa Mats, Corning
Supplier: Corning
Certificates
About this item
Silicone septa mat for capillary sequencing of 96-well PCR plates. Mat forms a tight seal on wells but is easily accessible through septa in each well. Ideal for work on ABI multi-capillary sequencing instruments.
Customers Who Bought This Also Bought