Graduated Midget Impingers, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Micro impinger designed for sampling of small volumes of air at low jet velocity, as per EPA Method 6
Impinger bottle is graduated from 5 to 30ml in 5ml subdivisions. Both the bottle and stopper have 24/40 standard taper joints. The orifice at the impinger tip is 1mm, passing 0.09 to 0.11 CFM of air at 12in H20 vacuum. Available with 12/5 ball joints or plain tubulations.
Customers Who Bought This Also Bought