A propos de cet article
Active clamp style current probe has dual sensitivity ranges, 500 mV/A and 50 mV/A for current measurements up to 7 A or 70 A, respectively. Bandwidth in both ranges up to 1 MHz. Powered by 9 V battery or included AC power adapter.
Features:
Compact stylish body
Dual sensitivity ranges
1 MHz bandwidth (-3 dB)
Over-range indicator
Accessories: Insulated BNC Cable, BNC to Banana Plugs Adapter, AC Power Adapter, and Instruction Manual. The series is marketed under the time-honored Elditest brand name in both North America and the European Union.
SPECIFICATIONS
Type: Active Current
Measurement Ranges: 500 mV/A / 50 mV/A
Current (DC + peak AC): 7 A / 70 A
DC Measurement Accuracy:
±3% ±20 mA @ 500 mV/A
(20 mA to 14 A peak)
±4% ±200 mA @ 50 mV/A
(200 mA to 100 A peak)
±15% max @ 50 mV/A
(100 A peak to 140 A peak)
Bandwidth: DC - 1 MHz
Rise and Fall Time: 0.35 µs (typ.)
Max. Working Voltage:
600 Vac RMS CAT II
300 Vac RMS CAT III
Max. Floating Voltage:
600 Vac RMS CAT II
300 Vac RMS CAT III
Battery Life: 6 hours
Operating Temperature/Humidity: 0°C to +50°C / 0% to 95%
Max. Cable Diameter:10.3 mm (0.4")
Dimensions: 262mm x 81mm x 36mm (10.3" x 3.2" x 1.4")
Weight: 310 g w/o battery
RoHS (2002/95/EC) Compliant
SPECIFICATIONS
Type: Active Current
Measurement Ranges: 500 mV/A / 50 mV/A
Current (DC + peak AC): 7 A / 70 A
DC Measurement Accuracy:
±3% ±20 mA @ 500 mV/A
(20 mA to 14 A peak)
±4% ±200 mA @ 50 mV/A
(200 mA to 100 A peak)
±15% max @ 50 mV/A
(100 A peak to 140 A peak)
Bandwidth: DC - 1 MHz
Rise and Fall Time: 0.35 µs (typ.)
Max. Working Voltage:
600 Vac RMS CAT II
300 Vac RMS CAT III
Max. Floating Voltage:
600 Vac RMS CAT II
300 Vac RMS CAT III
Battery Life: 6 hours
Operating Temperature/Humidity: 0°C to +50°C / 0% to 95%
Max. Cable Diameter:10.3 mm (0.4")
Dimensions: 262mm x 81mm x 36mm (10.3" x 3.2" x 1.4")
Weight: 310 g w/o battery
RoHS (2002/95/EC) Compliant