U-tek® Gel Pack Mats and Single Gel Pack Ice Packs, Sonoco ThermoSafe
Fournisseur: Sonoco Thermosafe
Certificats
A propos de cet article
U-TEK® refrigerant mats and singles remain frozen longer than regular ice mats without the melting mess.
- Three styles available
- Cushions fragile shipments
- May be refrozen and reused
- Unaffected by biocides
A convenient alternative to ice packs, single refrigerant packets are for use as a cubed ice substitute. Cut-to-size mats are for wrap-around applications. Single sheet pre-cut mats are also available. Mats cushion fragile shipments, and they won’t dilute ice baths or damage samples. May be refrozen and reused. Mats withstand pressures of 60 pounds per square inch for five minutes or longer. Unaffected by biocides, fungicides, and pest control agents. Refrigerant contains neutral pH distilled and deionized water and proprietary materials certified by the U.S. Food and Drug Administration as food grade chemicals.
Customers Who Bought This Also Bought