DISPOSAFLEX® Glass Columns, Kimble Chase
Fournisseur: Kimble Chase
Certificats
A propos de cet article
A multi-purpose, disposable glass column.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Supplied assembled with a polypropylene reservoir, a lower column fitting with a luer outlet and a 20 µm polyethylene bed support.
Customers Who Bought This Also Bought