To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:DyLight® 594
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:1.0 mg
- Concentration:1.0 mg/ml (E 1% at 280 nm = 13.0)
- Immunogen:Human IgG
- Purification:Affinity purified using solid phaseHuman IgG
- Storage buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2-8 °C
- Shipping temperature:2-8 °C
- Form:Lyophilized
- Format:Purified IgG
- Cross adsorption:No
- Tested applications:Fluoresence ELISA, Fluorescence-Activated Cell Sorting
- Cat. no.:10151-230
- Supplier No.:IR2576
Specifications
About this item
The ImmunoReagents' DyLight® 594 conjugates generate red fluorescence and shows wide excitation wavelength separation from green fluorophores for multi-color applications with reduced bleedthrough. The physical characteristics of DyLight® 594 are comparable to other 594 dyes. DyLight® 594 is available for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 593nm / 618nm Emission Color = Red (Similar Dyes: Alexa Fluor 594, Texas Red)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human