To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Goat
- Antigen symbol:IgG
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:1.0 mg
- Western blot:Yes
- Cross adsorption:Yes
- Format:Purified IgG
- Form:Clear, colorless liquid, 0.2 µm filtered
- Antibody fragment:F(ab')2
- Storage buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage temperature:2-8 °C
- Concentration:> 4.5 mg/ml (E 1% at 280 nm = 13.0)
- Shipping temperature:2-8 °C
- Immunogen:Goat IgG
- Tested applications:Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling
- Purification:Affinity purified using solid phase Goat IgG
- Cat. no.:10151-094
- Supplier No.:IR2488
Spécifications
A propos de cet article
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy. Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.
- Origin: US
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat