To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Rabbit
- Antigen symbol:IgG
- Conjugation:TRITC (Tetramethylrhodamine Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:0.5 mg
- Western blot:Yes
- Cross adsorption:No
- Format:Purified IgG
- Form:Lyophilized
- Antibody fragment:F(ab')2
- Storage buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2-8 °C
- Concentration:1.0 mg/ml (E 1% at 280 nm = 13.0)
- Shipping temperature:2-8 °C
- Immunogen:Rabbit IgG
- Tested applications:Immunomicroscopy
- Purification:Affinity purified using solid phaseRabbit IgG
- Cat. no.:10150-762
- Supplier No.:IR2234
Spécifications
A propos de cet article
TRITC (Tetramethylrhodamine isothiocyanate) is a popular rhodamine derivative pink in color but emits a red-orange color upon Emission at 580nm. TRITC is frequently paired with FITC in double labeling experiments and conjugated antibodies are often used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. These conjugates are best excited with the 532 nm spectral line of the He-Ne laser and are also recommended as a second color detection reagent in in situ hybridization applications. (Excitation/Emission = 555nm / 580nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, DyLight 550)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit