To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:DyLight® 633
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:0.5 mg
- Western blot:Yes
- Cross adsorption:No
- Format:Purified IgG
- Form:Lyophilized
- Antibody fragment:F(ab')2
- Storage buffer:10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
- Storage temperature:2-8 °C
- Concentration:1.0 mg/ml (E 1% at 280 nm = 13.0)
- Shipping temperature:2-8 °C
- Immunogen:Mouse IgG
- Tested applications:Fluoresence ELISA, Fluorescence-Activated Cell Sorting
- Purification:Affinity purified using solid phaseMouse IgG
- Cat. no.:10150-274
- Supplier No.:IR1934
Spécifications
A propos de cet article
The ImmunoReagents' DyLight® 633 conjugates fluoresces red with physical properties comparable to other 633 dyes. DyLight® 633 is ideal for multi-color applications. DyLight® 633 is available as reactive labeling agents and as conjugates of secondary antibodies and biotin-binding proteins for use in fluorescence microscopy, flow cytometry, Western blotting, ELISA, high-content screening and other array platforms. Excitation/Emission = 638nm / 658nm Emission Color = Red (Similar Dyes: Alexa Fluor 633)
- Origin: US
ImmunoReagents' antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.
Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse