Capillary Melting Point Tubes, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Tubes are manufactured from borosilicate glass and feature openings on both ends.
Tubes are packaged 100 per vial. The 0.25 mm thick tube conforms to U.S.P XXI requirements.
Customers Who Bought This Also Bought