Pregnancy Tests, Stanbio Laboratory
Fournisseur: Stanbio Laboratory
Certificats
A propos de cet article
Stanbio Laboratory offers pregnancy tests to meet your clinical and laboratory needs.
Sensivity in Urine 20 mIU/mL
Sensivity in Serum 10 mIU/mL (20 mIU/mL for the True® 20 Plus set)
Certifications: All pregnancy tests are FDA cleared and CE marked. Some tests have additional Health Canada certifications.
Ordering information: Tests are single step cassette platform formats for urine only or urine/serum pregnancy testing.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.