KIMBLE® FLEX-COLUMN® Sample Diffusion Disc, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
A transparent acrylic ring is sealed to a 20 μm porosity HDPE disc which protects the top of the gel bed when the sample is manually applied. A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.
Customers Who Bought This Also Bought