A propos de cet article
The Adventurer® AX Analytical balance series strike the ideal mix between inventive features, long-lasting durability, and uncomplicated weighing capabilities.
Stability, accuracy, and fast operation ensure optimal weighing results. This trio of characteristics ensures the most important aspects of routine laboratory weighing are covered: accurate results you can count on extremely quickly. The color touchscreen and connectivity options power a modern weighing experience. The wide viewing angle color touchscreen provides quick access to all of the unit’s applications, while dual USB ports with GLP/GMP tracking capabilities assist in the monitoring and reporting of data. A space-saving draftshield improves user experience and accessibility. The two-piece, top-mounted draftshield (mg and 0.1mg models only) doors open wide on both sides, providing ample room for sample placement. This also reduces the footprint of the draftshield in the rear of the balance when open.
The Adventurer® AX Analytical balance series strike the ideal mix between inventive features, long-lasting durability, and uncomplicated weighing capabilities.
The Adventurer® AX incorporates all of the features and applications necessary for simple weighing and also offers options for more complex measurements
The Adventurer® AX incorporates all of the features and applications necessary for simple weighing and also offers options for more complex measurements
Stability, accuracy, and fast operation ensure optimal weighing results. This trio of characteristics ensures the most important aspects of routine laboratory weighing are covered: accurate results you can count on extremely quickly. The color touchscreen and connectivity options power a modern weighing experience. The wide viewing angle color touchscreen provides quick access to all of the unit’s applications, while dual USB ports with GLP/GMP tracking capabilities assist in the monitoring and reporting of data. A space-saving draftshield improves user experience and accessibility. The two-piece, top-mounted draftshield (mg and 0.1mg models only) doors open wide on both sides, providing ample room for sample placement. This also reduces the footprint of the draftshield in the rear of the balance when open.
Operate and access the nine application modes and abundant features that eliminate the need to do several manual calculations through the modern color touchscreen. Operators can wear laboratory gloves while utilizing the touchscreen, eliminating the inconvenience and hazards associated with constantly putting on and removing gloves. In addition to the touchscreen, the Adventurer® AX also features six mechanical keys that provide tactile feedback and allow the operator to perform repetitive operations such as tare, zero, calibration, and print. All units feature a die-cast metal bottom housing, stainless steel pan, full housing replaceable in-use cover, and integral weigh below hook.
Applications include Weighing, Parts Counting, Percent Weighing, Check Weighing, Animal/Dynamic Weighing, Formulation, Totalization, Density Determination, and Display Hold.
Spécifications
- Catalog No:
- 10153-692
- 10153-694
- 10153-700
- 10153-702
- 10153-696
- 10153-698
- Operating temperature range:
- 10 to 30°C (50 to 86°F)
- 10 to 30°C (50 to 86°F)
- 10 to 30°C (50 to 86°F)
- 10 to 30°C (50 to 86°F)
- 10 to 30°C (50 to 86°F)
- 10 to 30°C (50 to 86°F)