VWR® Automatic Self-Zero Burets with Stopcocks, Class A, Serialized
Fournisseur: VWR International
Certificats
A propos de cet article
Made from borosilicate 3.3 expansion glass.
- Borosilicate glass fabrication
- Automatically self-zero
Buret assembly features a precision-grade white scale. Supplied with reservoir bottle, stopcock, and rubber bulb.
Ordering information: Burets come with individual certificate of compliance.
Spécifications
- Catalog No:
- 10124-014
- 10124-012
- 10124-010
- 10124-018
- 10124-016
- 10124-008
- Certification:
- Individual
- Individual
- Individual
- Individual
- Individual
- Individual