Rescue Critters® Squeekums Rat Manikin
# de catalogue 470126-198
Fournisseur: THALES & CO.
Spécifications
- Description:Squeekums Manikin
- Cat. no.:470126-198
- Supplier No.:320
Spécifications
A propos de cet article
Learn How To Handle Lab Rodents
- Practice handling lab rodents
- Students can practice iv access at the tail and ear tagging procedures
- Made to order
The tail of this amazing rodent model can be conveniently detached and disposed of, while also having the capability of IV access at the caudal vein site. Tiny replaceable ears help students learn ear tagging procedures. The model is fully articulated and extremely realistic. Head, feet, and limbs move in a natural manner.
Squeekums comes with its own protective case, IV accessories, artificial blood, and instructions.
Ordering information: This item is made to order. Returns cannot be accepted.
Customers Who Bought This Also Bought