Rescue Critters® K-9 I.V. Trainer
# de catalogue 470126-194
Fournisseur: THALES & CO.
Spécifications
- Description:K-9 IV Trainer
- Cat. no.:470126-194
- Supplier No.:404
Spécifications
A propos de cet article
Veterinary IV Trainer
- Practice injections and blood draws
- Made to order
Specifically built to teach the drawing of blood and performing of injections, the Canine IV Trainer is the perfect pedagogical tool for this purpose. It is realistically sculpted as the hind limb of a dog. This simple product will help students become accustomed to these tactile procedures.
Ordering information: This item is made to order. Returns cannot be accepted.
Customers Who Bought This Also Bought