VWR® Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
Fournisseur: VWR International
Certificats
A propos de cet article
Made from borosilicate 3.3 expansion glass.
- Borosilicate glass construction
- Individually serialized
- Glass penny head stopper
- Amber glass recommended for UV light-sensitive samples
Flasks are serialized and supplied with glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought