VWR® Wide Mouth Volumetric Flasks with PE Stoppers, Class A, Serialized
Fournisseur: VWR International
Certificats
A propos de cet article
Made from borosilicate 3.3 expansion glass.
- Borosilicate glass construction
- Individually serialized
- Provided with polyethylene stopper
- Wide-mouthed design
Flasks are serialized and supplied with polyethylene stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought