VWR® Micro Volumetric Flasks with Glass Stoppers, Class A, Serialized
Supplier: VWR International
Certificates
About this item
Clear glass volumetric flasks are calibrated to contain at 20°C and comply to ASTM E237 standards.
- Clear siding
Flasks are seralized and supplied with one glass penny head stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought