VWR® Kohlrausch Volumetric Mud Flasks, Class A, Serialized
Supplier: VWR International
Certificates
About this item
Kohlrausch flasks feature a marking spot and an enlarged upper neck to permit easy introduction of solids.
- Made from Borosilicate 3.3 expansion glass
- Calibrated to contain at 20°C
Kolhraush Flasks are made from Borosilicate 3.3 expansion glass.
Ordering information: Flasks are individually certified.
Customers Who Bought This Also Bought