Impulse Heat Sealer, PPC Flexible Packaging
Supplier: Fisher Container
Certificates
About this item
The model 216X Impulse Heat Sealer produces 40.6 cm (16") seals with a maximum sealing thickness of 0.16"
The 210X Impulse Heat Sealer produces 20.3 cm (8") seals. These units are ideal for cleanroom or laboratory use in film or bag sealing applications. The Impulse Heat Sealer is supplied with two element covers and two pressure pad covers.
Customers Who Bought This Also Bought