KIMBLE® CHROMAFLEX® Chromatography Column Polypropylene Ferrule Fittings, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Polypropylene ferrule fitting used to make connections between the tubing and the end fittings of CHROMAFLEX® columns.
Customers Who Bought This Also Bought