Bag Sealer, A Daigger
Supplier: A Daigger
Certificates
About this item
Bag Sealers are designed to create seals on plastic sheets, bags, or flat tubing
Safely seal polyethylene and polypropylene plastics up to 5mil thick. These 115V units feature a timing knob for safe sealing and a 1.8m cord.
Customers Who Bought This Also Bought