A propos de cet article
The Boekel Scientific Microplate Shaker features a low profile and small footprint and are available in a 2 Plate or 4 Plate Model
A simple screw attachment allows for quick and easy fitting of any standard depth 96 or 384 well plate while keeping your microplates securely in place. The LCD Display with microprocessor controls provide for easy viewing in the lab and includes a count down timer with auto shut off which emits an audible end of cycle alarm. This unit is also ideal for use in an incubator and cold rooms.
All Boekel Scientific products come with a 2-year warranty.
Each model features microprocessor controls to provide a regulated, uniform shaking while the LCD display is easily viewed in the lab. With a speed range of 250 to 1000 RPM's and a 2mm orbit, this unit is ideal for immunoassays or general microplate shaking.
Spécifications
- Catalog No:
- 97004-388
- 97004-386
- Speed range:
- 250-1000 rpm
- 250-1000 rpm
- Amplitude orbitale (mm):
- 2 mm
- 2 mm
- Orbit:
- 2 mm
- 2 mm
- Dimensions:
- 7W x 7.25D x 3.5H"
- 7W x 7.25D x 3.5H"
- Timer:
- 1 minute to 96 hours (with auto shut off)
- 1 minute to 96 hours (with auto shut off)
- Electrical:
- 12V DC via 115/230V AC Adapter
- 12V DC via 115/230V AC Adapter