VWR® Mini Hotplate Stirrer, 120 V
Catalog # 10153-308
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Temperature range:to 375°C
- Description:VWR® Mini Hotplate Stirrer
- Overall dimensions:20L×15.2W×12.4H cm (7.9×6×4.9")
- Speed range:100 - 1200 rpm
- Electrical:120V, 50/60Hz
- Shipping weight:1.8 kg (4 lbs.)
- Cat. no.:10153-308
- Maximum Capacity:1000 mL
Specifications
About this item
Rugged, compact hotplate stirrers feature an ergonomic design and are ideal for educational labs.
- Rugged 5x5" Aluminum Top Plate
- Boils 300 mL of Water in 16 minutes
- Built-In Support Rod Holder
- Five-Year Warranty on Parts and Labor
These units can heat and stir up to 1000 mL of water
The built-in support rod holder with locking knob accepts an optional support rod and clamp kit. Kit includes 45.7 cm (18”) support rod, thermometer extension clamp, three-prong medium swivel clamp, and hook connector. Optional support plate allows hot plate to be mounted to lab frames or ring stands.
Durable 5x5" cast aluminum top plate will not chip or crack, and provides a uniform heating surface. Bi-metallic thermostat offers reliable temperature control. Powerful heater reaches maximum temperature in minutes. Powerful motor and magnet deliver reliable and consistent stirring.
Certifications: TUV listed. CE marked. RoHS certified.
Ordering information: The 120V units include a 183 cm (72"), 3-wire cord and plug. All units include a five-year limited warranty on parts and labor.
Customers Who Bought This Also Bought