3B Scientific® Skin
# de catalogue 470103-258
Fournisseur: American 3B Scientific
Spécifications
- Description:Skin
- Height:16 cm
- No. of Keyed Items:31
- Cat. no.:470103-258
- Length:35 cm
- Width:40 cm
- Supplier No.:J16
- No. of Parts:3
Spécifications
A propos de cet article
- Buy individually or on a single backer
- Plaque mounted and can be viewed on a table, or hung
- Three years manufacturers warranty
- Includes 3B smart anatomy
The model consists of three individual parts on a common stand that represent sections of the human skin with a magnification of 80x. Anatomical differences of the skin sections are didactically represented in an understandable manner. All layers making up the skin and microscopic structures such as nerve, vessels and tactile corpuscles have been accurately and spatially realized on the models.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought