3B Scientific® Shoulder Joint With Rotator Cuff
# de catalogue 470103-190
Fournisseur: American 3B Scientific
Spécifications
- Description:Shoulder Joint With Rotator Cuff
- Cat. no.:470103-190
- Supplier No.:A880
- No. of Parts:5
Spécifications
A propos de cet article
This shoulder joint model consists of the upper half of the humerus, as well as the clavicle and scapula.
- Show the structure of the knee in sections
- 3 year manufacturers warranty
- Includes 3B smart anatomy
Besides showing the musculature of the rotator cuff, the shoulder joint model also shows the origin and insertion points of the shoulder muscles highlighted in color (origin = red, insertion points = blue). Upon detaching the individual muscles, all movements of the shoulder joint can be carried out.
BodyTextAccessories: For information about 3B smart anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought