3B Scientific® Skull With Facial Muscles
# de catalogue 470102-960
Fournisseur: American 3B Scientific
Spécifications
- Description:Skull With Facial Muscles
- Height:22 cm
- No. of Keyed Items:42
- Cat. no.:470102-960
- Length:16 cm
- Width:16 cm
- Supplier No.:A300
- No. of Parts:1
Spécifications
A propos de cet article
- Study The Early Bone Growth Of The Spinal Column
- Includes Stand
- 3 Year Manufacturers Warranty
- Includes 3B Smart Anatomy
The face and mastication muscles are illustrated on the right half of this skull model. The face musculature can easily and precisely be differentiated from the mastication musculature by using two colors. On the left half the muscle origins and insertions are marked with colors (origin: red, insertion: blue). The jaw is movable and due to the flexible musculature the rudimentary chewing motion can be demonstrated. Cranium and m. masseter are detachable.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought