Polyethylene Stoppers, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Low cost polyethylene stoppers are an alternative to standard glass and PTFE stoppers
Leak-proof, air-tight seals and wide top flange for ease of use.
Customers Who Bought This Also Bought