VWR® Men's Lab Coat, Reusable
Fournisseur: VWR International
Certificats
A propos de cet article
Classic white lab coats for men are manufactured from durable and comfortable poly/cotton (80/20 blend) fabric for long-term wear
- Durable, comfortable poly/cotton (80/20 blend) fabric
- Plain front, no embroidery
- Industrial launderable
Overall length is 99cm (39"). Available in white only.
These industrial launderable garments feature a quick-release five-snap front, one breast pocket with a pencil slot, and two oversized hip pockets (large enough for a mini-tablet). Additionally, there are side vents for easy pants pocket access.
Ordering information: Sizes XX-Small, X-Small, 4X-Large, and 5X-Large are also available for special order. Contact your Avantor sales representative for more information. Please allow up to four weeks for delivery.
Spécifications
- Catalog No:
- 10141-302
- 10141-304
- 10141-300
- 10141-306
- 10141-308
- 10141-310
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- Polyester/cotton (80/20 blend) fabric
- Polyester/cotton (80/20 blend) fabric
- Polyester/cotton (80/20 blend) fabric
- Polyester/cotton (80/20 blend) fabric
- Polyester/cotton (80/20 blend) fabric
- Polyester/cotton (80/20 blend) fabric
Spécifications
Informations connexes
Customers Who Bought This Also Bought