Syringe Needle Flask Adapters, Kimble Chase
# de catalogue KT197800-2440
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cap Type:Blind Hole Rubber Stopper
- Cat. no.:KT197800-2440
- ST Joint Size:[ST] 24/40
- Supplier No.:197800-2440
Spécifications
A propos de cet article
Provides access to flasks and other containers when pierced by a syringe needle
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
When used in conjunction with standard syringes, liquid samples may be added or withdrawn from multiple neck flasks. Allows syringe needles to be used as capillary bleed tubes or bubblers.
Ordering information: Supplied with one Blind Hole Rubber Stopper fitted to the top.
Customers Who Bought This Also Bought