Spécifications
- Description:Fluorescent Total Protein Stain for 1D and 2D Gels
- Volume:5 ml
- Supplier No.:AC2124
- Sufficient For:20 Minigels
Spécifications
A propos de cet article
AzureRed Fluorescent Protein Stain is a quantitative total protein stain for blots and blots with the sensitivity to detect less than 1 ng of protein per band or spot
Gels and blots stained with AzureRed can be imaged on any fluorescent imaging system, including laser- and CCD-based systems.
Fully compatible with downstream Western blotting or mass spectrometry, AzureRed is non-toxic and biodegradable, for safe and simple disposal.