Level Sensors, S.C.A.T.
Fournisseur: Avantor
Certificats
A propos de cet article
These level sensors provide dependable protection from overflow when filling barrels. The float positions itself securely at the opening by its own weight while the barrel fills up.
- For barrel opening with Ø 50 to 90 mm
- Practical handhold
- Easy to use with gloves
PFA float, PTFE/PE indicator
These level sensors are a reliable protection against overflow during the filling.
Customers Who Bought This Also Bought