Coliwasa Tubes, Kimble Chase, DWK Life Sciences
# de catalogue 56925-225
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Cat. no.:56925-225
- O.D.×L:22×1067 mm (0.875×42")
- Supplier No.:76100-200
Spécifications
A propos de cet article
The Coliwasa tube is used to obtain a vertical column of liquid representing an accurate cross-section of the sampled material
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Two-part borosilicate glass tube has a ball in the lower end which acts as a check valve to prevent sample loss during transfer. Both parts are prescored, allowing the unit to be easily snapped apart for safe disposal.
Customers Who Bought This Also Bought