Specifications
- Volume:1000 µl (1 ml)
- Cat. no.:KT748019-0001
- Graduations:0.01 cc
- Supplier No.:748019-0001
Specifications
About this item
The Plastic Luer Slip Micro Syringes work for chromatography, nuclear medicine, HPLC sample preparation, and many laboratory procedures.
- Two Part
- Plastic
- Polypropylene Barrel
- Polyethylene Plunger
- Latex Free
More chemically resistant than rubber-tipped syringes, these two-part plastic syringes feature a polypropylene barrel and polyethylene, low dead-space plunger
The needles are for research use only and are not for use in diagnostic procedures.
The sterile, individually wrapped needles contain no rubber, silicone oil, styrene, DNA, latex, or DEHP.