Adhesive Crystallography Seal, Thermo Scientific
Catalog # 89499-576
Supplier: Thermo Fisher Scientific
Specifications
- Optical property:Optically clear
- Temperature Range:–70 to 110°C (–94 to 230°F)
- Description:Adhesive Crystallography Seal Roll
- Cat. no.:89499-576
- Supplier No.:AB-2450
Specifications
About this item
Optically clear, pressure sensitive, non-sticky adhesive seal for crystallography and genotyping applications.
Samples are not exposed to adhesive. Seal is easily peelable without leaving residue on the plate. Rolls are designed for use with the Thermo Scientific SEAL-IT 100™ adhesive sealer.
Customers Who Bought This Also Bought