About this item
RPMI 1640 is suitable for a wide range of anchorage independent cells. RPMI 1640 Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS). Although, RPMI-1640 has been shown to support the serum-free growth of a number of cells.
- Endotoxin and Mycoplasma tested
RPMI 1640 is suitable for a wide range of anchorage independent cells
RPMI-1640 medium was developed by Moore et al., at Roswell Park Memorial Institute, hence the acronym RPMI. The formulation is based on the RPMI-1630 series of media utilizing a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins. RPMI-1640 medium has been used for the culture of human normal and neoplastic leukocytes. RPMI-1640 when properly supplemented, has demonstrated wide applicability for supporting growth of many types of cell cultures.
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). Although, RPMI-1640 has been shown to support the serum-free growth of a number of cells.
Common applications of RPMI-1640 include: fusion protocols and growth of hybrid cells, culture of normal human leukocytes in monolayer and suspension, and support of HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinoma growth (to name a few).
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 Medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). Although, RPMI-1640 has been shown to support the serum-free growth of a number of cells.
The original Click formulation was published in 1972 and is a modification of Eagle's Minimum Essential Medium with Hanks' salts (HMEM). RPMI 1640: Click;s Medium (1:1) has increased concentrations of many essential amino acids, addition of non-essential amino acids, sodium pyruvate and nucleic acid precursors.
Wilson & Horster used the 1:1 mix of RPMI-1640 and Click's medium to study the differential response of defined distal nephron epithelia in culture. RPMI-1640 Click's medium is frequently used for the suspension, washing and culture of T-cells