ContactUsLinkComponent
Glutamine XL 200 mMr, Quality Biological
Glutamine XL 200 mMr, Quality Biological
Catalog # 10128-832
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Glutamine XL 200 mMr, Quality Biological
Catalog # 10128-832
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Glutamine XL, 200 mM
  • Size:
    100 mL
  • Storage conditions:
    2°C to 8°C
  • Sterility:
    Sterile Filtered
  • Shipping conditions:
    Ambient temperature
  • Test description:
    Endotoxin and Performance tested
  • Contents:
    Stabilized L-glutamine
  • Storage:
     2 to 8° C
  • Shipping temperature:
    Ambient
  • Cat. no.:
    10128-832
  • Shelf Life:
    24 months
  • Supplier No.:
    118-288-061
Specifications
About this item
Glutamine XL is useful as a cell culture media supplement
  • With stable L-Alanyl-Glutamine
Standard, unstabilized solutions of L-glutamine spontaneously break down over time to release cytotoxic amines. Glutamine XL is a stabilized solution that prevents degradation and cytotoxicity, with the benefit of a long shelf life.
The incorporated L-alanine protects the unstable alpha amino acid group of L-glutamine. Aminopeptidase within the cells cleaves the dipeptide, gradually releasing both L-glutamine and L- alanine for use.
Recommendation
Dilute Glutamine XL the same way as L-glutamine in the appropriate cell culture medium. Typically, L-glutamine is diluted to concentration of 1-4 mM. Protect from light.
Sterile Filtered
Endotoxin and Mycoplasma Tested
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured in an ISO 9001:2015 certifed company in accordance with ISO-14644 and IEST-RP-006.3; meeting standards for ISO Class 8 and ISO Class 7.
Caution: Research Use Only
Frequently Bought Together