Splitter, Reflux/Distillation Adapter, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
The Reflux/Distillation Splitter is used to allow easy switching of the distilate path by means of adjusting the valve position to either open or closed.
- Compact design
- Integrated Swagelok takeoff side arm
- Streamlined vacuum-assisted distilate transfers
This in-line adapter simplifies the vapor flow path, and it's compact design and integrated Swagelok takeoff side arm make vacuum assisted distilate transfers streamlined. The splitters are available in both a straight or angled configuration. Angled adapters are used with 10° angled side necks and transition to a vertical 90° position.
Customers Who Bought This Also Bought