Drum Samplers, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Conventional open-end drum thieves provide a quick method for sampling unmarked drums
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Units are easily broken for quick disposal.
Customers Who Bought This Also Bought