Educational-Grade Burets, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
These economical and versatile burets are ideal for use in educational institutions and for many general laboratory procedures.
- Funnel fill style
- Easy-to-read durable black enamel scale
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The 17021H burets are supplied with a PTFE compression fit stopcock assembly and a precision ground tip. The 17040K buret is supplied with a PTFE stopcock plug.
Ordering information: The KIM-KAP™ dust cap is included.
Customers Who Bought This Also Bought