Solution Bottle with Narrow Mouth for Rubber Stopper, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Ideal for storage and dispensing of solutions.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Neck is tooled for increased mechanical strength and for a uniform fit with rubber stoppers.
Customers Who Bought This Also Bought