Reservoir Bottle with Bottom Valve Outlet, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Designed to store and discharge liquids via a bottom valve outlet.
KIMAX® bottles are graduated with an approximate capacity scale. Bottom outlet is controlled with an 8 mm valve which can be used with ¹/₄" flexible tubing attached to the quick-release hose barb connector and with rigid ¹/₄" Ext.Ø PTFE tubing with the o-ring and cap supplied on the quick-release connector.
Certifications: Ref: ASTM Method D1744. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought