Conical Thin-Wall Micro Reaction Tubes with Attached Caps, Bio Plas
Fournisseur: Bio Plas
Certificats
A propos de cet article
Conical Thin-Wall Micro Reaction Tubes are fabricated from virgin polypropylene and feature highly polished inner surfaces to allow for maximum sample recovery
The Multi-Cap Tool assists in providing secure lid closings and openings, and prevents aerosolizing.
Tubes also featuer an 0.5 ml frosted flat cap surface for sample inscription. Tubes are autoclavable at 121 °C (250 °F), 15psi, for fifteen minutes.
Customers Who Bought This Also Bought